Merck
Hyperaromatic stabilization of arenium ions.
Organic letters (2010-11-09)
Jaya S Kudavalli, Derek R Boyd, Dara Coyne, James R Keeffe, David A Lawlor, Aoife C MacCormac, Rory A More O'Ferrall, S Nagaraja Rao, Narain D Sharma
ABSTRACT
Benzene-cis- and trans-1,2-dihydrodiols undergo acid-catalyzed dehydration at remarkably different rates: k(cis)/k(trans) = 4500. This is explained by formation of a β-hydroxycarbocation intermediate in different initial conformations, one of which is stabilized by hyperconjugation amplified by an aromatic no-bond resonance structure (HOC(6)H(6)(+) ↔ HOC(6)H(5) H(+)). MP2 calculations and an unfavorable effect of benzoannelation on benzenium ion stability, implied by pK(R) measurements of -2.3, -8.0, and -11.9 for benzenium, 1-naphthalenium, and 9-phenanthrenium ions, respectively, support the explanation.
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
Sigma-Aldrich
1,4-Cyclohexadiene, 97%