Merck
  • 2-Phenoxyethanol derived diselenide and related compounds; synthesis of a seven-membered seleninate ester.
2-Phenoxyethanol derived diselenide and related compounds; synthesis of a seven-membered seleninate ester.
Organic & biomolecular chemistry (2010-11-05)
Santosh K Tripathi, Sagar Sharma, Harkesh B Singh, Ray J Butcher
ABSTRACT
Syntheses of several diorganodiselenides and, in particular, a seven-membered cyclic seleninate ester derived from 2-phenoxyethanol are described. The seleninate ester was obtained from allyl (2-(2-hydroxyethoxy)phenyl) selenide through a series of oxidation and [2,3] sigmatropic rearrangement steps. The ester exhibits good GPx-like activity in the coupled reductase assay.
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard