Merck
  • Partitioning behavior of an acid-cleavable, 1,3-dioxolane alkyl ethoxylate, surfactant in single and binary surfactant mixtures for 2- and 3-phase microemulsion systems according to ethoxylate head group size.
Partitioning behavior of an acid-cleavable, 1,3-dioxolane alkyl ethoxylate, surfactant in single and binary surfactant mixtures for 2- and 3-phase microemulsion systems according to ethoxylate head group size.
Journal of colloid and interface science (2010-10-01)
Javier Gomez del Rio, Douglas G Hayes, Volker S Urban
ABSTRACT
Partition coefficients for a pH-degradable 1,3-dioxolane alkyl ethoxylate surfactant, 4-CH(3)O (CH(2)CH(2)O)(5.6)-CH(2), 2,2-(CH(2))(12)CH(3), 2-(CH(2)) CH(3), 1,3-dioxolane or "cyclic ketal" surfactant, CK-2,13-E(5.6,ave), between isooctane- and water-rich phases of 2- and 3-phase microemulsion systems (K(n)) were determined as functions of the ethoxylate size, n, and temperature for the neat surfactant and its binary surfactant mixtures, to understand the partitioning of alkyl ethoxylates possessing a broad distribution of ethoxylate size and to determine conditions required for formation of 3-phase microemulsion systems at an optimal temperature where phase separation occurs rapidly, important for protein purification via proteins' selective partitioning to the middle phase, driven by affinity to the second surfactant of the binary mixture. A semi-empirical thermodynamic mathematical model described the partitioning data well, provided optimal temperature values consistent with phase diagrams and theory, and demonstrated that the tail region of CK-2,13-E(5.6,ave) is more polar than the hydrophobes of fatty alcohol ethoxylates. The addition of Aerosol-OT (AOT) removed the temperature sensitivity of CK-2,13-E(5.6,ave)s partitioning, producing 3-phase microemulsion systems between 20 °C and 40 °C. Analysis of the bottom phases of the 2- and 3-phase microemulsion systems formed by CK-2,13-E(5.6,ave) via small-angle neutron scattering demonstrated the presence of spherical, monodisperse oil-in-water microemulsions.
Product Description
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
2,2,4-Trimethylpentane, analytical standard
Sigma-Aldrich
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
Sigma-Aldrich
1,3-Dioxolane, ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%
Sigma-Aldrich
2,2,4-Trimethylpentane, suitable for HPLC, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Sigma-Aldrich
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
1,3-Dioxolane, anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%