Merck
  • [Markers of chloric acid (I) in biological systems--identification and properties].
[Markers of chloric acid (I) in biological systems--identification and properties].
Postepy biochemii (2010-09-29)
Agnieszka Robaszkiewicz
ABSTRACT
Reactive oxygen and nitrogen species are attributed to initiation and propagation of many diseases. The demonstration of elevated activity of myeloperoxidase and the level of 3-chlorotyrosine in atherosclerosis, kidney diseases and chronic inflammations brought about the interest in the biological role of another strong oxidant--hypochlorite. Concentration of this compound is extremely difficult to estimate in vivo and in vitro because of its high reactivity. The reaction of hypochlorite with biological compounds lead to formation of chlorohydrins, glutathione sulfonamides, chloramines, 3- and 3,5-dichlorotyrosines and chlorinated DNA bases (8-chloroadenine, 8-chloroguanine, 5-chlorocytosine and 5-chlorouracil). At least some of these products of hypochlorite action are believed to provide specific HOCl-biomarkers, useful especially in the analysis of clinical samples, using sensitive detection techniques.
Sigma-Aldrich
Potassium chlorate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chlorate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium chlorate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium chlorate, puriss., 99-101%
Millipore
Potassium chlorate Supplement, suitable for microbiology