Merck
  • Microwave-assisted synthesis and reversed-phase high-performance liquid chromatographic separation of diastereomers of (R,S)-baclofen using ten chiral derivatizing reagents designed from trichloro-s-triazine.
Microwave-assisted synthesis and reversed-phase high-performance liquid chromatographic separation of diastereomers of (R,S)-baclofen using ten chiral derivatizing reagents designed from trichloro-s-triazine.
Journal of chromatography. A (2010-09-08)
Ravi Bhushan, Shuchi Dixit
ABSTRACT
Four dichloro-s-triazine (DCT) and five monochloro-s-triazine (MCT) chiral derivatizing reagents (CDRs) were synthesized by incorporating amino acid amide moieties as chiral auxiliaries in trichloro-s-triazine and its 6-methoxy derivative, respectively. Another MCT reagent was synthesized by substitution of two chlorine atoms with two different amino acid amides in trichloro-s-triazine. These reagents were used for synthesis of diastereomers of (R,S)-baclofen under microwave irradiation (i.e. 60 s at 85% power using DCT reagents and 90 s at 85% power using MCT reagents). The diastereomers were separated on a reversed-phase C18 column using mixtures of methanol with aqueous trifluoroacetic acid (TFA) with UV detection at 230 nm. The separation behavior in terms of retention times and resolutions obtained for the two sets of diastereomers prepared with DCT and MCT reagents were compared among themselves and among the two groups. Longer retention times and better resolutions were observed with DCT reagents as compared to MCT reagents. The calibration curves were linear for both (R)- and (S)-baclofen in the concentration range 50-500 μg/ml. The average regression was 0.999 for both (R)- and (S)-baclofen. The RSD for (R)-baclofen was 0.40-0.86% for intra-day precision and 0.60-1.40% for inter-day precision and these values for (S)-baclofen were 0.52-0.75% and 0.64-1.32%, respectively. The recovery was 97.2-98.9% for (R)- and 97.0-98.9% for (S)-baclofen. The limit of detection was 1.63 ng/ml and 1.52 ng/ml for (R)- and (S)-baclofen, respectively.
Sigma-Aldrich
Cyanuric chloride, 99%