Merck
  • Modular amino acids-based chiral ligands for copper-catalyzed enantioselective conjugation addition of diethylzinc to cyclic enones.
Modular amino acids-based chiral ligands for copper-catalyzed enantioselective conjugation addition of diethylzinc to cyclic enones.
Chirality (2010-06-16)
Shaohua Gou, Zaher M A Judeh
ABSTRACT
New amino acid-based modular chiral ligands were readily synthesized and used to catalyze the asymmetric conjugate addition of Et(2) Zn to various cyclic enones in the presence of a variety of copper sources. Moderately high ee of up to 72% were obtained using ligand (S)-1e under mild conditions.
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes