Merck
  • Destruction of acenaphthene, fluorene, anthracene and pyrene by a dc gliding arc plasma reactor.
Destruction of acenaphthene, fluorene, anthracene and pyrene by a dc gliding arc plasma reactor.
Journal of hazardous materials (2010-05-14)
Liang Yu, Xin Tu, Xiaodong Li, Yu Wang, Yong Chi, Jianhua Yan
ABSTRACT
In this study, four kinds of PAHs (polycyclic aromatic hydrocarbons) i.e. acenaphthene, fluorene, anthracene and pyrene are used as targets for investigation of PAHs treatment process assisted by dc gliding arc discharge. The effects of carrier gas and external resistance on the PAHs decomposition process are discussed. The results indicate that the destruction rate can be achieved to the highest with the carrier gas of oxygen and the external resistance of 50 kOmega independent of type of PAHs. Furthermore, experimental results suggest that destruction energy efficiency of gliding arc plasma would be improved by treating higher concentration pollutants. Based on the analysis of experimental results, possible destruction mechanisms in different gas discharge are discussed.
Product Description
Supelco
Acenaphthene, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acenaphthene, 99%
Pricing and availability is not currently available.
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.