Merck
  • Microbial conversion of rare ginsenoside Rf to 20(S)-protopanaxatriol by Aspergillus niger.
Microbial conversion of rare ginsenoside Rf to 20(S)-protopanaxatriol by Aspergillus niger.
Bioscience, biotechnology, and biochemistry (2010-01-09)
Lei Liu, Li-Juan Gu, Dong-Liang Zhang, Zhen Wang, Chun-Yan Wang, Zheng Li, Chang-Keun Sung
ABSTRACT
In this study, rare ginsenoside Rf was transformed into 20(S)-protopanaxatriol (PPT(S)) by glycosidase from Aspergillus niger. By investing the reaction conditions, the optimal conditions were obtained, as follows: pH 5.0, temperature 55 degrees C, and substrate concentration 1.25 mmol/l. Under optimal conditions, PPT(S) (1.13 micromol) prepared from 1.25 mumol Rf showed a higher yield (90.4%). The enzymatic reaction was analyzed by reversed-phase HPLC, suggesting the transformation pathway: Rf-->Rh1(S)-->PPT(S).
Product Description
Supelco
Ginsenoside Rf, analytical standard
Ginsenoside Rf, primary reference standard