Merck
  • Unprecedented marriage of a cationic pentanuclear cluster and a 2D polymeric anionic layer based on a flexible tripodal ligand and a Cu(II) ion.
Unprecedented marriage of a cationic pentanuclear cluster and a 2D polymeric anionic layer based on a flexible tripodal ligand and a Cu(II) ion.
Inorganic chemistry (2010-01-06)
Zheng-Bo Han, Guo-Xin Zhang, Ming-Hua Zeng, Da-Qiang Yuan, Qian-Rong Fang, Jian-Rong Li, Joan Ribas, Hong-Cai Zhou
ABSTRACT
The anionic CdI(2)-type topological net, [Cu(2)(tci)(2)](2-), and the pentanuclear copper cluster cation [Cu(5)(tci)(2)(OH)(2)(H(2)O)(8)](2+) [tci = tris(2-carboxyethyl)isocyanurate] form a complementary 3D supramolecular framework. Interestingly, there exist centrosymmetric cyclic (H(2)O)(18) clusters in the cavities.
Sigma-Aldrich
Cadmium iodide, 99%
Sigma-Aldrich
Cadmium iodide, purum p.a., ≥99.0% (T)