Merck
  • Vibrational spectroscopy of secondary amine salts: 1. Assignment of NH(2)(+) stretching frequencies in crystalline phases.
Vibrational spectroscopy of secondary amine salts: 1. Assignment of NH(2)(+) stretching frequencies in crystalline phases.
The journal of physical chemistry. B (2009-11-13)
Guinevere A Giffin, Scott Boesch, Dharshani N Bopege, Douglas R Powell, Ralph A Wheeler, Roger Frech
ABSTRACT
The NH(2)(+) stretching modes of secondary amine salts have been previously studied, but the band assignments are inconsistent between the various studies. This paper assigns characteristic NH(2)(+) group frequencies between approximately 2500 and 2400 cm(-1). Crystal structures of four diamine salts are reported here. Vibrational frequencies were calculated with the B3LYP hybrid Hartree-Fock/density functional method and the 6-31G(d) split-valence plus polarization basis set, and the results are in agreement with the experimental frequencies. Deuterium dilution experiments result in a group of sharply featured bands between the NH(2)(+) and the ND(2)(+) stretching bands. These bands, located between 2200 and 2100 cm(-1), are attributed to modes that contain contributions from coupled N-H and N-D stretching motions.
Sigma-Aldrich
Hydrogen bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%