Merck
  • A concise synthesis of castanospermine by the use of a transannular cyclization.
A concise synthesis of castanospermine by the use of a transannular cyclization.
The Journal of organic chemistry (2009-10-28)
Thomas Jensen, Mette Mikkelsen, Anne Lauritsen, Thomas L Andresen, Charlotte H Gotfredsen, Robert Madsen
ABSTRACT
A nine-step synthesis of (+)-castanospermine has been accomplished in 22% overall yield from methyl alpha-D-glucopyranoside. The key transformations involve a zinc-mediated fragmentation of benzyl-protected methyl 6-iodoglucopyranoside, ring-closing olefin metathesis, and strain-release transannular cyclization to afford the indolizidine skeleton of the natural product.
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%