Merck
Evidence-based review of clinical studies on pulpectomy.
Journal of endodontics (2009-07-28)
ABSTRACT
In this section, we analyze recent studies on pulpectomy procedures. These procedures represent an interesting example of dental treatment offered by general dentists, pediatric dentists, and endodontists. Accordingly, there is a need for a centralized source of analysis of these studies. It is our belief that this special issue might serve that function.
Sigma-Aldrich
Iodoform, 99%