Merck
  • Copper(II) complexes of coumarin-derived Schiff bases and their anti-Candida activity.
Copper(II) complexes of coumarin-derived Schiff bases and their anti-Candida activity.
Journal of inorganic biochemistry (2009-07-28)
Bernadette S Creaven, Michael Devereux, Dariusz Karcz, Andrew Kellett, Malachy McCann, Andy Noble, Maureen Walsh
ABSTRACT
The condensation of 7-amino-4-methyl-coumarin (1) with a number of substituted salicylaldehydes yielded a series of Schiff bases (2a-2k) in good yields. Subsequent reaction of these ligands with copper(II) acetate yielded Cu(II) complexes (3a-3k) and some were characterised using X-ray crystallography. All of the free ligands and their metal complexes were tested for their anti-Candida activity. A number of the ligands and complexes exhibited anti-Candida activity comparable to that of the commercially available antifungal drugs, ketoconazole and Amphotericin B.
Sigma-Aldrich
7-Amino-4-methylcoumarin, Chromophore for substrates
Pricing and availability is not currently available.
Sigma-Aldrich
7-Amino-4-methylcoumarin, 99%
Pricing and availability is not currently available.