Merck
  • Cyclic oligomer of oxide clusters through a siloxane bond. Synthesis and structure of reaction products of alpha2-mono-lacunary Dawson polyoxometalate with tetrachlorosilane and tetraethoxysilane.
Cyclic oligomer of oxide clusters through a siloxane bond. Synthesis and structure of reaction products of alpha2-mono-lacunary Dawson polyoxometalate with tetrachlorosilane and tetraethoxysilane.
Dalton transactions (Cambridge, England : 2003) (2009-07-10)
Takayuki Kurashina, Shotaro Aoki, Ryota Hirasawa, Takeshi Hasegawa, Yuhki Kasahara, Shoko Yoshida, Kenji Yoza, Kenji Nomiya
ABSTRACT
The title compound with the formula (Me2NH2)13H5[{alpha2-P2W17O61(Si2O)}3(micro-O)3].39H2O Me2NH(2)-1 was obtained in 12.8% (0.258 g scale) yield by a 1:2 molar ratio reaction of alpha2-mono-lacunary Dawson polyoxometalate (POM) [alpha2-P2W17O61]10- with SiCl4 in water and unequivocally characterized by complete elemental analysis, TG/DTA, FTIR, (solution and solid-state) 31P NMR, solid-state 29Si NMR and X-ray crystallography. [Note: the moieties of the polyoxoanions in Me2NH(2)-1 and Me2NH(2)-2 are abbreviated simply as 1 and 2, respectively.] X-Ray crystallography revealed that the molecular structure of polyoxoanion 1 was a cyclic trimer consisting of three alpha2-mono-lacunary Dawson POM subunits constituted through intra- and intermolecular siloxane bonds. The compound resulted in an approximate Cs symmetry, but not the idealized C3v symmetry, because one Dawson unit was unequivalently incorporated into the cyclic framework. This compound is the first example of a siloxane-bonding cyclic trimer of POMs and its formation suggests that the oxygen atoms in the lacunary site of POM are reactive just like the surface oxygen atoms of silica. The formation of the cyclic trimer was also clarified by two control experiments: (1) the reaction of [alpha2-P2W17O61]10- with Si(OEt)4 under acidic conditions (pH 1), and (2) oligomerization under 6 M aqueous HCl acidic conditions (pH approximately 0) of the reactive monomer units (RMUs: tentatively assigned as "[alpha2-P2W17O61{O(SiOH)2}]6-"), derived from the cleavage of Ph-Si bonds by the thermal degradation of [alpha2-P2W17O61{O(SiPh)2}]6-. On the other hand, the reactions of alpha2-mono-lacunary Dawson POMs with GeX4 (X=OEt, Cl) gave the monomeric species of mono-germanium-substituted Dawson POM (Me2NH2)7[alpha2-P2W17O61(GeOH)].5H2O Me2NH(2)-2, which was characterized by CHN analysis, TG/DTA, FTIR, X-ray crystallography, and (solid-state and solution) 31P NMR spectroscopy. X-ray crystallography revealed that compound Me2NH(2)-2 was a monomeric species, but the Ge site per formula unit was not determined due to disorder of alpha-Dawson structure.
Sigma-Aldrich
Silicon tetrachloride, 99.998% trace metals basis
Sigma-Aldrich
Silicon tetrachloride, 99%
Sigma-Aldrich
Silicon tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Silicon tetrachloride solution, 1.0 M in methylene chloride