Merck
  • Amphiphilic polyesters derived from silylated and germylated fatty compounds.
Amphiphilic polyesters derived from silylated and germylated fatty compounds.
Biomacromolecules (2009-03-20)
Nadia Katir, Abdelkrim El Kadib, Mohamed Dahrouch, Annie Castel, Nicolas Gatica, Zahra Benmaarouf, Pierre Riviere
ABSTRACT
New classes of amphiphilic polyesters were prepared from metallated (Si, Ge) fatty methyl ester (FAME) precursors and poly(tetramethylene oxide) glycol. Hydrosilylation of 10-undecenoic methyl ester by tetramethyldisiloxane occurred at 80 degrees C in the presence of Karstedt's catalyst, and hydrogermylation of the same FAME derivative was obtained at the same temperature under radical AIBN initiation. These diester precursors, obtained in high yields (approximately 90%), reacted with poly(tetramethylene oxide) glycol under free solvent to give silicon polymers or germanium oligomers. These condensed materials display both the characteristic of organic-inorganic hybrid materials and those of amphiphilic polymers. The nature of organometallic fragment (hydrophobicity of tetramethyldisiloxy and sterical hindrance of diphenylgermyl) was shown to influence the chemical reactivity of the polymerizable monomers and the physical properties of the resulting copolymers. The amphiphilicity of these materials provides a driving force for the formation of small objects (approximately 1 nm), making them very attractive as hybrid nanocontainers.
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00
Sigma-Aldrich
Poly(tetrahydrofuran), average Mn ~2,900
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00
Sigma-Aldrich
10-Undecynoic acid, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00
Sigma-Aldrich
Poly(tetrahydrofuran), average Mn ~1,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00
Sigma-Aldrich
Poly(tetrahydrofuran), average Mn ~2,000, contains BHT as stabilizer
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
PLN 315.00