Merck
  • Sorption mechanisms of cephapirin, a veterinary antibiotic, onto quartz and feldspar minerals as detected by Raman spectroscopy.
Sorption mechanisms of cephapirin, a veterinary antibiotic, onto quartz and feldspar minerals as detected by Raman spectroscopy.
Environmental pollution (Barking, Essex : 1987) (2009-02-28)
Jonathan W Peterson, Theresa A O'Meara, Michael D Seymour, Wei Wang, Baohua Gu
ABSTRACT
Raman spectroscopy was used to investigate sorption mechanisms of cephapirin (CHP), a veterinary antibiotic, onto quartz (SiO(2)) and feldspar (KAlSi(3)O(8)) at different pH. Sorption occurs by electrostatic attraction, monodentate and bidentate complexation. The zwitterion (CHP(o)) adsorbs to a quartz((+)) surface by electrostatic attraction of the carboxylate anion group (-COO(-)) at low pH, but adsorbs to a quartz((-)) surface through electrostatic attraction of the pyridinium cation, and possibly COO(-) bridge complexes, at higher pH. CHP(-) bonds to quartz((-)) surfaces by bidentate complexation between one oxygen of -COO(-) and oxygen from carbonyl of an acetoxymethyl group. On a feldspar((+/-)) surface, CHP(o) forms monodentate complexes between CO, and possible -COO(-) bridges and/or electrostatic attachments to localized edge (hydr)oxy-Al surfaces. CHP(-) adsorbs to feldspar((-)) through monodentate CO complexation. Similar mechanisms may operate for other cephalosporins. Results demonstrate, for the first time, that Raman techniques can be effective for evaluating sorption mechanisms of antibiotics.
Product Description
Supelco
Cefapirin sodium, analytical standard