- A stable hyponitrite-bridged iron porphyrin complex.
A stable hyponitrite-bridged iron porphyrin complex.
Journal of the American Chemical Society (2009-02-05)
Nan Xu, Adam L O Campbell, Douglas R Powell, Jana Khandogin, George B Richter-Addo
PMID19191487
ABSTRACT
The coupling of two nitric oxide (NO) molecules in heme active sites is an important contributor to the conversion of NO to nitrous oxide (N(2)O) by heme-containing enzymes. Several formulations for the presumed heme-Fe{N(2)O(2)}(n-) intermediates have been proposed previously, however, no crystal structures of heme-Fe{N(2)O(2)}(n-) systems have been reported to date. We report the first isolation and characterization of a stable bimetallic hyponitrite iron porphyrin, [(OEP)Fe](2)(mu-N(2)O(2)), prepared from the reaction of [(OEP)Fe](2)(mu-O) with hyponitrous acid. Density functional theoretical calculations were performed on the model compound [(porphine)Fe](2)(mu-N(2)O(2)) to characterize its electronic structure and properties.
MATERIALS