Merck
  • Molecular recognition and self-assembly special feature: Encapsulation and characterization of proton-bound amine homodimers in a water-soluble, self-assembled supramolecular host.
Molecular recognition and self-assembly special feature: Encapsulation and characterization of proton-bound amine homodimers in a water-soluble, self-assembled supramolecular host.
Proceedings of the National Academy of Sciences of the United States of America (2009-02-03)
Michael D Pluth, Dorothea Fiedler, Jeffrey S Mugridge, Robert G Bergman, Kenneth N Raymond
ABSTRACT
Cyclic amines can be encapsulated in a water-soluble self-assembled supramolecular host upon protonation. The hydrogen-bonding ability of the cyclic amines, as well as the reduced degrees of rotational freedom, allows for the formation of proton-bound homodimers inside of the assembly that are otherwise not observable in aqueous solution. The generality of homodimer formation was explored with small N-alkyl aziridines, azetidines, pyrrolidines, and piperidines. Proton-bound homodimer formation is observed for N-alkylaziridines (R = methyl, isopropyl, tert-butyl), N-alkylazetidines (R = isopropyl, tert-butyl), and N-methylpyrrolidine. At high concentration, formation of a proton-bound homotrimer is observed in the case of N-methylaziridine. The homodimers stay intact inside the assembly over a large concentration range, thereby suggesting cooperative encapsulation. Both G3(MP2)B3 and G3B3 calculations of the proton-bound homodimers were used to investigate the enthalpy of the hydrogen bond in the proton-bound homodimers and suggest that the enthalpic gain upon formation of the proton-bound homodimers may drive guest encapsulation.
Sigma-Aldrich
N-Methylpyrrolidine, 97%
Sigma-Aldrich
1-Methylpyrrolidine, ≥98.0% (GC)