Merck
  • Characterization of a ring-hydroxylating dioxygenase from phenanthrene-degrading Sphingomonas sp. strain LH128 able to oxidize benz[a]anthracene.
Characterization of a ring-hydroxylating dioxygenase from phenanthrene-degrading Sphingomonas sp. strain LH128 able to oxidize benz[a]anthracene.
Applied microbiology and biotechnology (2009-01-28)
Luc Schuler, Yves Jouanneau, Sinéad M Ní Chadhain, Christine Meyer, Maria Pouli, Gerben J Zylstra, Pascal Hols, Spiros N Agathos
ABSTRACT
Sphingomonas sp. strain LH128 was isolated from a polycyclic aromatic hydrocarbon (PAH)-contaminated soil using phenanthrene as the sole source of carbon and energy. A dioxygenase complex, phnA1fA2f, encoding the alpha and beta subunit of a terminal dioxygenase responsible for the initial attack on PAHs, was identified and isolated from this strain. PhnA1f showed 98%, 78%, and 78% identity to the alpha subunit of PAH dioxygenase from Novosphingobium aromaticivorans strain F199, Sphingomonas sp. strain CHY-1, and Sphingobium yanoikuyae strain B1, respectively. When overexpressed in Escherichia coli, PhnA1fA2f was able to oxidize low-molecular-weight PAHs, chlorinated biphenyls, dibenzo-p-dioxin, and the high-molecular-weight PAHs benz[a]anthracene, chrysene, and pyrene. The action of PhnA1fA2f on benz[a]anthracene produced two benz[a]anthracene dihydrodiols.
Product Description
Benz[a]anthracene, BCR®, certified reference material
Sigma-Aldrich
Benz[a]anthracene, 99%
Supelco
Benz[a]anthracene, analytical standard
Supelco
Benz[a]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland