Merck
  • Hydrothermal synthesis of Ni(12)P(5) hollow microspheres, characterization and photocatalytic degradation property.
Hydrothermal synthesis of Ni(12)P(5) hollow microspheres, characterization and photocatalytic degradation property.
Journal of colloid and interface science (2009-01-16)
Jun Li, Yonghong Ni, Kaiming Liao, Jianming Hong
ABSTRACT
In this paper, we report the successful synthesis of Ni(12)P(5) hollow spheres via a facile hydrothermal route, employing white phosphorus (WP) and nickel nitrate as the reactants in the presence of hexamethylenetetramine (HMT) and polyethylene glycol 10000 (PEG-10000). The phase and morphology of the product were characterized by means of powder X-ray diffraction (XRD), energy dispersive spectrometry (EDS), transmission electron microscopy (TEM), and scanning electron microscopy (SEM). HMT and surfactant (PEG-10000) played important roles in the formation of Ni(12)P(5) hollow microspheres. Furthermore, research also showed that the as-prepared Ni(12)P(5) hollow spheres could photocatalytically degrade some organic dyes such as Safranine T and Pyronine B under irradiation of 365 nm UV light.
Sigma-Aldrich
Polyethylene glycol, 10000, for synthesis
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, purum p.a., crystallized, ≥97.0% (KT)
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, puriss. p.a., ≥98.5% (KT)
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, crystals