Merck
A facile strategy for nonenzymatic glucose detection.
Analytical biochemistry (2008-11-27)
Yinling Wang, Dandan Zhang, Weiwei Zhang, Feng Gao, Lun Wang
ABSTRACT
Glassy carbon electrode modified with boron oxide nanoparticles supported on multiwall carbon nanotubes was obtained via a facile approach. The as-prepared modified electrode exhibits excellent electrocatalytic activity toward the redox of glucose in pH 7.0 phosphate buffer solution. The electrochemical response of the modified electrode to glucose shows a linear range of 1.5-260 microM with a correlation coefficient of 0.9986 and the calculated detection limit is 0.8 microM at a signal-to-noise ratio of 3, which makes it useful for developing the electrochemical determination of glucose concentrations without using glucose oxidase at physiological pH.
Sigma-Aldrich
Boric anhydride, puriss. p.a., ≥98% (T)
Sigma-Aldrich
Boric anhydride, 99.999% trace metals basis
Sigma-Aldrich
Boric anhydride, granulated, ≥98.0% (T)
Supelco
Boric anhydride, 99.98% trace metals basis