Merck
  • "Ligand-free" CuI-catalyzed highly efficient intramolecular S-vinylation of thiols with vinyl chlorides and bromides.
"Ligand-free" CuI-catalyzed highly efficient intramolecular S-vinylation of thiols with vinyl chlorides and bromides.
The Journal of organic chemistry (2008-11-27)
Qiwu Zhao, Ling Li, Yewen Fang, Deqian Sun, Chaozhong Li
ABSTRACT
With CuI as the catalyst and K(3)PO(4) x 3 H(2)O as the base, highly efficient intramolecular S-vinylation of thiols with vinyl chlorides or bromides was successfully implemented without the help of an additional ligand. Moreover, the competition experiments revealed that the 4-exo cyclization is fundamentally preferred over other modes (5-exo, 6-exo, and 6-endo) of cyclization.
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF
Sigma-Aldrich
Vinyl bromide, 98%