Merck
  • Detection of high-charge density polyanion contaminants in biomedical heparin preparations using potentiometric polyanion sensors.
Detection of high-charge density polyanion contaminants in biomedical heparin preparations using potentiometric polyanion sensors.
Analytical chemistry (2008-11-15)
Lin Wang, Stacey Buchanan, Mark E Meyerhoff
ABSTRACT
A very simple and inexpensive method to detect oversulfated chondroitin sulfate (OSCS) or other high-charge density polyanionic structures as contaminants in heparin products using potentiometric polyanion sensors is described. In the potentiometric measurement, a greater change in the phase boundary equilibrium potential of polymeric membranes formulated with tridodecylmethylammonium (TDMA) anion exchange sites is observed for the contaminated heparin than for the untainted heparin due to the higher charge density of OSCS or other impurities compared to that of porcine heparin. Detection of 0.5 wt % OSCS impurity is readily achieved using only 1 mg/mL of final polyion concentration. Even lower detection limits for OSCS contamination may be possible if higher final concentrations of heparin preparations can be employed in the test procedure.
Sigma-Aldrich
Tridodecylmethylammonium iodide, 97%
Supelco
Tridodecylmethylammonium chloride, Selectophore
Sigma-Aldrich
Tridodecylmethylammonium chloride, 98%