Merck
  • Asymmetric cycloadditions of o-quinone methides employing chiral ammonium fluoride precatalysts.
Asymmetric cycloadditions of o-quinone methides employing chiral ammonium fluoride precatalysts.
Organic letters (2008-10-15)
Ethan Alden-Danforth, Michael T Scerba, Thomas Lectka
ABSTRACT
The catalytic, enantioselective, [4 + 2] cycloaddition reaction of ortho-quinone methides with silyl ketene acetals is described. This mechanistically interesting reaction, initiated by a chiral cinchona alkaloid-derived ammonium fluoride "precatalyst" complex, affords a variety of alkyl- and aryl-substituted 3,4-dihydrocoumarin products in excellent yield and with good enantioselectivity.
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis