- Fluorination reactions in microreactors.
Fluorination reactions in microreactors.
Chemical communications (Cambridge, England) (2008-08-09)
Tomas Gustafsson, Ryan Gilmour, Peter H Seeberger
PMID18688335
ABSTRACT
The DAST-mediated conversion of a range of alcohols to the corresponding fluorides in a microstructured device is described. This safe, practical fluorination method will facilitate reactions currently challenging on large scale.