Merck
Positive patch-test reactions to platinum are rare.
Dermatitis : contact, atopic, occupational, drug (2008-07-17)
Joseph F Fowler, Jennifer H Perryman, Branden Quinlan
ABSTRACT
Although contact allergy to some metals is common, patch testing for allergy to platinum is rarely performed. To determine the prevalence of positive patch-test reactions to platinum in an unselected consecutive group of patients presenting for diagnostic patch testing. Unselected patients who presented with a possible diagnosis of contact dermatitis were tested with a series of allergens, including ammonium tetrachloroplatinate 0.25% aqueous. Of 446 patients tested, only 2 females showed positive reactions to platinum; one had no other positive reactions, and the other had reactions to other metals. Positive patch-test reactions to platinum are rare. However, platinum jewelry may occasionally provoke allergic contact dermatitis.
Sigma-Aldrich
Ammonium tetrachloroplatinate(II), 99%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 98%
Sigma-Aldrich
Potassium tetrachloroplatinate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloroplatinate(II), ≥99.9% trace metals basis