Merck
  • Determination of aflatoxins B1, B2, G1, and G2 and ochratoxin A in ginseng and ginger by multitoxin immunoaffinity column cleanup and liquid chromatographic quantitation: collaborative study.
Determination of aflatoxins B1, B2, G1, and G2 and ochratoxin A in ginseng and ginger by multitoxin immunoaffinity column cleanup and liquid chromatographic quantitation: collaborative study.
Journal of AOAC International (2008-06-24)
Mary W Trucksess, Carol M Weaver, Carolyn J Oles, Frederick S Fry, Gregory O Noonan, Joseph M Betz, Jeanne I Rader
ABSTRACT
The accuracy, repeatability, and reproducibility characteristics of a method using multitoxin immunoaffinity column cleanup with liquid chromatography (LC) for determination of aflatoxins (AF; sum of aflatoxins B1, B2, G1, and G2) and ochratoxin A (OTA) in powdered ginseng and ginger have been established in a collaborative study involving 13 laboratories from 7 countries. Blind duplicate samples of blank, spiked (AF and OTA added) at levels ranging from 0.25 to 16.0 microg/kg for AF and 0.25 to 8.0 microg/kg for OTA were analyzed. A naturally contaminated powdered ginger sample was also included. Test samples were extracted with methanol and 0.5% aqueous sodium hydrogen carbonate solution (700 + 300, v/v). The extract was centrifuged, diluted with phosphate buffer (PB), filtered, and applied to an immunoaffinity column containing antibodies specific for AF and OTA. After washing the column with water, the toxins were eluted from the column with methanol, and quantified by high-performance LC with fluorescence detection. Average recoveries of AF from ginseng and ginger ranged from 70 to 87% (at spiking levels ranging from 2 to 16 microg/kg), and of OTA, from 86 to 113% (at spiking levels ranging from 1 to 8 microg/kg). Relative standard deviations for within-laboratory repeatability (RSDr) ranged from 2.6 to 8.3% for AF, and from 2.5 to 10.7% for OTA. Relative standard deviations for between-laboratory reproducibility (RSDR) ranged from 5.7 to 28.6% for AF, and from 5.5 to 10.7% for OTA. HorRat values were < or = 2 for the multi-analytes in the 2 matrixes.
Sigma-Aldrich
Aflatoxin B2
Supelco
Aflatoxin B2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin G2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Aflatoxin B2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B2
Aflatoxin G2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin G2
Sigma-Aldrich
Aflatoxin G1, from Aspergillus flavus
Sigma-Aldrich
Aflatoxin G2