Merck
  • FT-IR and theoretical study of 3,5-dimethyl-1H-pyrazole-1-carboxamidine (L) and the complexes CoL2(H2O)2(NO3)2, NiL2(H2O)2(NO3)2.
FT-IR and theoretical study of 3,5-dimethyl-1H-pyrazole-1-carboxamidine (L) and the complexes CoL2(H2O)2(NO3)2, NiL2(H2O)2(NO3)2.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-06-20)
Peter Pogány, Attila Kovács, Katalin Mészáros Szécsényi, Vukadin M Leovac
ABSTRACT
In the paper a joint experimental and theoretical study of 3,5-dimethyl-1H-pyrazole-1-carboxamidine (L) as well as its complexes CoL2(H2O)2(NO3)2 and NiL2(H2O)2(NO3)2 is reported. On the basis of FT-IR experiments and a DFT-derived scaled quantum mechanical force field the normal coordinate analysis of L was carried out. The FT-IR spectra of the two complexes were interpreted using the present assignment of L and computed vibrational data of the complexes. The ionic and charge transfer interactions in the complexes were assessed by means of natural bond orbital (NBO) analysis.
Sigma-Aldrich
3,5-Dimethyl-1-pyrazolylformaminidium nitrate, 97%