Merck
Carbohydrate functionalization using cationic iron carbonyl complexes.
Carbohydrate research (2008-05-13)
Anders Bergh, Henrik Gradén, Núria Parera Pera, Thomas Olsson, Ulf J Nilsson, Nina Kann
ABSTRACT
Cationic iron carbonyl cyclohexadiene complexes were employed in the derivatization of the 3-OH position of unprotected and protected methyl beta-D-galactopyranosides using two different approaches, giving access to galactopyranosides with an aromatic or cyclohexadienoic functionality in this position.
Sigma-Aldrich
Iron(0) pentacarbonyl
Sigma-Aldrich
Iron(0) pentacarbonyl, >99.99% trace metals basis