- Optimized pyroelectric properties of 0-3 composites of PZT particles in polyurethane doped with lithium perchlorate.
Optimized pyroelectric properties of 0-3 composites of PZT particles in polyurethane doped with lithium perchlorate.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control (2008-02-16)
Bernd Ploss, Markus Krause
PMID18276541
ABSTRACT
A substantial improvement in the performance of pyroelectric 0-3 composites of ceramic particles in a polymer matrix has been achieved by doping the polymer matrix material. Readily prepared and polarized films with various volume fractions of lead zirconate-titanate (PZT) particles in polyurethane have been doped in a solution of lithium perchlorate in acetone to increase the conductivity. With an appropriate conductivity, the dielectric permittivities of the ceramic particles and the polymer matrix become matched, resulting in an improvement of the pyroelectric coefficient from about 6 microC/(m(2)K) to about 50 microC/(m(2)K). The experimental results are explained by theoretical predictions.