Merck
  • Electron-rich heteroaroylphosphonates and their reaction with trimethyl phosphite.
Electron-rich heteroaroylphosphonates and their reaction with trimethyl phosphite.
Organic & biomolecular chemistry (2008-01-26)
D Vaughan Griffiths, Mohamad J Al-Jeboori, Yuen-Ki Cheong, Philip Duncanson, Jayne E Harris, Michael C Salt, Helen V Taylor
ABSTRACT
Dialkyl heteroaroylphosphonates based on thiophene, pyrrole or furan have been prepared and their reactions with trimethyl phosphite investigated. Deoxygenation of the carbonyl groups in these heteroaroylphosphonates occurs to give carbene intermediates, which then undergo further reaction. In the case of the furan-3-oylphosphonates and those systems containing a thiophene or pyrrole ring, the major reaction pathway involves intermolecular trapping of the carbene intermediates by the trimethyl phosphite, leading to the formation of ylidic phosphonates that can be readily converted into the corresponding 1,1-bisphosphonates. However, in some furan-2-oylphosphonates the carbenes generated undergo ring-opening to initially give acyclic alkynylphosphonates which may react further to give other novel phosphorus compounds. The effects of substituents on the extent to which intermolecular trapping of the initially formed carbene competes with intramolecular rearrangement has been investigated. The latter process appears to be suppressed by a substituent at the 5-position of the furan ring, the resulting ylidic phosphonates being a rare example of an efficient intermolecular trapping of a furan-2-yl carbene.
Sigma-Aldrich
Trimethyl phosphite, ≥97%
Sigma-Aldrich
Trimethyl phosphite, ≥99%