Merck
  • N-heterocyclic carbene ligands in copper-catalyzed addition of diethylzinc to N-sulfonylimines.
N-heterocyclic carbene ligands in copper-catalyzed addition of diethylzinc to N-sulfonylimines.
Chemical & pharmaceutical bulletin (2008-01-08)
Abu Bakar Md, Yumiko Suzuki, Masayuki Sato
ABSTRACT
N-Heterocyclic carbenes (NHCs) were generated in-situ from imidazolium and imidazolinium salts by deprotonation of C-2 hydrogen and were used as ligands in the copper-catalyzed addition of diethylzinc to N-sulfonylimines. The copper-NHC complexes were shown to possess an efficient ligand acceleration effect (LAE).
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes