Merck
  • Synthesis of modified nucleosides for incorporation of formyletheno and carboxyetheno adducts of adenine nucleosides into oligonucleotides.
Synthesis of modified nucleosides for incorporation of formyletheno and carboxyetheno adducts of adenine nucleosides into oligonucleotides.
Nucleosides, nucleotides & nucleic acids (2007-12-07)
Niangoran Koissi, Harri Lönnberg
ABSTRACT
Three protected derivatives of 1,N(6)-ethenoadenine nucleosides, viz. 3-[5-O-(4,4'-dimethoxytrityl) of 7-formyl-(1) and 7-(1,2-diacetyloxypropyl)-2'-deoxyadenosine (2), and 3-[5-O-(4,4'-dimethoxytrityl)-2-O-(tert-butyldimethylsilyl)-7-(ethoxycarbonyl)adenosine (3), expected to allow introduction of formyletheno and carboxyethenoadenine adducts into oligonucleotides by the conventional phosphoramidite chemistry, have been synthesized.
Sigma-Aldrich
Ethyl bromoacetate, purum, ≥97.0% (GC)
Sigma-Aldrich
Ethyl bromoacetate, reagent grade, 98%