Merck
  • The O-acylation of ketone enolates by allyl 1H-imidazole-1-carboxylate mediated with boron trifluoride etherate: a convenient procedure for the synthesis of substituted allyl enol carbonates.
The O-acylation of ketone enolates by allyl 1H-imidazole-1-carboxylate mediated with boron trifluoride etherate: a convenient procedure for the synthesis of substituted allyl enol carbonates.
The Journal of organic chemistry (2007-10-30)
Barry M Trost, Jiayi Xu
ABSTRACT
A convenient access to substituted allyl enol carbonates was established through the reaction of ketone enolates with the complex of allyl 1H-imidazole-1-carboxylates and boron trifluoride etherate.
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis
Sigma-Aldrich
Boron trifluoride diethyl etherate, for synthesis
Sigma-Aldrich
2-Methyl-1-indanone, 99%