Merck
Criss-cross cycloadditions on ketazines derived from alicyclic ketones.
Molecules (Basel, Switzerland) (2007-10-27)
Jirí Verner, Milan Potácek
ABSTRACT
The reactivity of alicyclic ketazines in criss-cross cycloadditions was investigated. They react with potassium cyanate and ammonium thiocyanate in the presence of acetic acid to form spirocyclic perhydro[1,2,4]triazolo[1,2-alpha][1,2,4]triazole-1,5-diones and perhydro[1,2,4]triazolo[1,2-alpha][1,2,4]triazole-1,5-dithiones, respectively, in relatively high yields.
Sigma-Aldrich
Potassium cyanate, 96%
Sigma-Aldrich
Potassium cyanate-15N, ≥98 atom % 15N, ≥95% (CP)