Merck
  • Adiponectin suppresses IkappaB kinase activation induced by tumor necrosis factor-alpha or high glucose in endothelial cells: role of cAMP and AMP kinase signaling.
Adiponectin suppresses IkappaB kinase activation induced by tumor necrosis factor-alpha or high glucose in endothelial cells: role of cAMP and AMP kinase signaling.
American journal of physiology. Endocrinology and metabolism (2007-10-18)
Xiangdong Wu, Kalyankar Mahadev, Lauren Fuchsel, Raogo Ouedraogo, Shi-Qiong Xu, Barry J Goldstein
ABSTRACT
Adiponectin is a protein secreted from adipocytes that exhibits salutary effects in the vascular endothelium by signaling mechanisms that are not well understood. In obesity-related disease states and type 2 diabetes, circulating substances, including tumor necrosis factor-alpha (TNFalpha) and high glucose, activate IkappaB kinase (IKK)beta and reduce the abundance of its substrate, inhibitor of kappaB (IkappaB)alpha, leading to nuclear translocation of the transcription factor NF-kappaB and stimulation of an inflammatory signaling cascade closely associated with endothelial dysfunction. The present study demonstrates that the globular domain of adiponectin (gAd) potently suppresses the activation of IKKbeta by either TNFalpha or high glucose in human umbilical vein endothelial cells and ameliorates the associated loss of IkappaBalpha protein. Interestingly, activation of AMP kinase was substantially more effective than cAMP signaling in suppressing high glucose-induced IKKbeta activity, whereas both pathways were comparably active in suppressing the TNFalpha-induced increase in IKKbeta. Both cAMP/protein kinase A signaling and activation of the AMP kinase pathway played a role in the suppression by gAd of TNFalpha- and high glucose-mediated IKKbeta activation. These findings support an important role for adiponectin in anti-inflammatory signaling in the endothelium and also imply that multiple pathways are involved in the cellular effects of adiponectin.
Sigma-Aldrich
2′,5′-Dideoxyadenosine, ≥95% (HPLC), solid