Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of drip application of ammonium thiosulfate on fumigant degradation in soil columns.
Effect of drip application of ammonium thiosulfate on fumigant degradation in soil columns.
Journal of agricultural and food chemistry (2007-09-13)
Ruijun Qin, Suduan Gao, Bradley D Hanson, Jason A McDonald, Thomas J Trout, Husein Ajwa
ABSTRACT
Low permeability tarps can effectively minimize fumigant emissions while improving fumigation efficacy by retaining fumigants under the tarp. However, when planting holes are cut through the tarps, high-concentration fumigants may be released and result in environmental and worker safety hazards. In a 11-day column study, we explored the effect of drip irrigation application of ammonium thiosulfate (ATS) on 1,3-dichloropropene (1,3-D) and chloropicrin (CP) degradation in soil. Decrease of 1,3-D and CP concentrations in soil-gas phase followed a three-parameter logistic equation for all treatments. It was slowest in the control with a half-life ( t 1/2) of 86.0 h for 1,3-D and of 16.3 h for CP and most rapid when ATS was applied at 4:1 ATS/fumigant molar ratio with a half-life of 9.5 h for 1,3-D and of 5.5 h for CP. Our results indicate that applying ATS via the drip-irrigation systems to soil can accelerate fumigant degradation in soil and thus reduce emissions. This technical solution may be applicable in raised-bed strawberry production where drip-application of fumigants under tarps has become common.
Sigma-Aldrich
Ammonium thiosulfate, 98%