Merck
  • Novel polymer-supported chiral catalysts for the asymmetric addition of diethylzinc to aldehydes.
Novel polymer-supported chiral catalysts for the asymmetric addition of diethylzinc to aldehydes.
Chemical & pharmaceutical bulletin (2007-07-03)
Su-Xi Wang, Fen-Er Chen
ABSTRACT
Newly synthesized polymer-supported chiral amino alcohol catalysts 5a and 5b have been proved to be effective for the enantioselective addition of diethylzinc to aldehydes, affording the corresponding sec-alcohols in moderate enantiomeric excesses with good to excellent yields. The recycled catalyst could be reused in the same reaction without significant deterioration in performance.
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene