Merck
  • Novel multicomponent reaction of [60]fullerene: the first example of 1,4-dipolar cycloaddition reaction in fullerene chemistry.
Novel multicomponent reaction of [60]fullerene: the first example of 1,4-dipolar cycloaddition reaction in fullerene chemistry.
Organic & biomolecular chemistry (2007-02-23)
Guan-Wu Wang, Jia-Xing Li
ABSTRACT
The novel multicomponent reaction of [60]fullerene with dimethyl acetylenedicarboxylate and isoquinoline, quinoline or substituted quinolines was investigated. This type of reaction presents the first example of a 1,4-dipolar cycloaddition reaction in fullerene chemistry.
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%