- Honeybee Apis mellifera acetylcholinesterase--a biomarker to detect deltamethrin exposure.
Honeybee Apis mellifera acetylcholinesterase--a biomarker to detect deltamethrin exposure.
The purpose of this study is to investigate the possibility to use acetylcholinesterase (AChE) as a biomarker of exposure to deltamethrin insecticide in the honeybee, Apis mellifera and to test its reliability in the presence of other contaminants, as carbamate insecticide. Joined actions of deltamethrin (pyrethroid) and pirimicarb (carbamate), alone or in association, are investigated on AChE activity in surviving and dead honeybees, with a special focus on the relative proportions of its membrane and soluble forms. At the 0.5X dose (12.5 ng of deltamethrin and/or 2.5 microg of pirimicarb per bee), the residual tissue AChE activity in dead bees was 78% with deltamethrin, 43% with pirimicarb and 33% with dual treatment. In surviving bees, tissue AChE activity represented 250%, and 270% of control AChE activity with deltamethrin and dual treatment, respectively. The analysis of membrane and soluble AChE forms revealed an increase in the soluble form in dead bees after deltamethrin and dual treatment. However, in vitro investigations showed no direct interaction of deltamethrin on soluble and membrane AChE activity. The results suggest that the action of deltamethrin on AChE activity, in honeybee intact organisms, could be due to indirect mechanisms. The duality of AChE response to deltamethrin exposure, exhibited by the possibility of increase (surviving bees) or decrease (dead bees) of its activity has been pointed out for the first time. The important increase in AChE activity in response to deltamethrin, not altered by pirimicarb treatment, suggests that AChE activity could represent a robust biomarker specific to deltamethrin exposure in living bees.