Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Improvement in isolation and identification of food-derived peptides in human plasma based on precolumn derivatization of peptides with phenyl isothiocyanate.
Improvement in isolation and identification of food-derived peptides in human plasma based on precolumn derivatization of peptides with phenyl isothiocyanate.
Journal of agricultural and food chemistry (2006-07-20)
Misako Aito-Inoue, Kozo Ohtsuki, Yasushi Nakamura, Eun Young Park, Koji Iwai, Fumiki Morimatsu, Kenji Sato
ABSTRACT
For the isolation and detection of food-derived peptides in blood, an approach based on the derivatization of peptides with phenyl isothiocyanate (PITC) was developed. This approach allows hydrophilic peptides to be resolved and specifically detected by reversed-phase (RP) HPLC. For the rapid capturing and clarification of peptides in human plasma, solid-phase extraction by using a mini spin column (5 mmx5 mm) packed with a strong cation exchanger was used. The clarified peptide fraction was further fractionated by size-exclusion chromatography (SEC). The peptides in the SEC fractions were derivatized with PITC, and the derivatives were resolved by RP-HPLC by using an ammonium acetate buffer or a trifluoroacetic acid system. An automatic peptide sequencer based on Edman degradation with a modified program can directly analyze the resolved derivatives. Some synthetic peptides and food-derived peptides in human plasma were successfully isolated and identified by this approach.
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%