Merck
  • A cold active (2R,3R)-(-)-di-O-benzoyl-tartrate hydrolyzing esterase from Rhodotorula mucilaginosa.
A cold active (2R,3R)-(-)-di-O-benzoyl-tartrate hydrolyzing esterase from Rhodotorula mucilaginosa.
Applied microbiology and biotechnology (2006-07-19)
Christian Zimmer, Tanja Platz, Neza Cadez, Friedrich Giffhorn, Gert-Wieland Kohring
ABSTRACT
In a screening procedure a pink-colored yeast was isolated from enrichment cultures with (2R,3R)-(-)-di-O-benzoyl-tartrate (benzoyl-tartrate) as the sole carbon source. The organism saar1 was identified by morphological, physiological, and 18S ribosomal DNA/internal transcribed spacer analysis as Rhodotorula mucilaginosa, a basidiomycetous yeast. During growth the yeast hydrolyzed the dibenzoyl ester stoichiometrically to the monoester using the separated benzoate as the growth substrate, before the monoester was further cleaved into benzoate and tartrate, which were both metabolized. The corresponding benzoyl esterase was purified from the culture supernatant and characterized as a monomeric glycosylated 86-kDa protein with an optimum pH of 7.5 and an optimum temperature of 45 degrees C. At 0 degrees C the esterase still exhibited 20% of the corresponding activity at 30 degrees C, which correlates it to psychrophilic enzymes. The esterase could hydrolyze short chain p-nitrophenyl-alkyl esters and several benzoyl esters like benzoyl-methyl ester, ethylene-glycol-dibenzoyl ester, phenyl-benzoyl ester, cocaine, and 1,5-anhydro-D: -fructose-tribenzoyl ester. However feruloyl-ethyl ester was not hydrolyzed. The activity characteristics let the enzyme appear as a promising tool for synthesis of benzoylated compounds for pharmaceutical, cosmetic, or fine chemical applications, even at low temperatures.
Sigma-Aldrich
(+)-2,3-Dibenzoyl-D-tartaric acid, ≥98%, made from synthetic tartaric acid
Sigma-Aldrich
(+)-2,3-Dibenzoyl-D-tartaric acid, ≥99.0% (T)
Sigma-Aldrich
Dibenzoyl-L-tartaric acid, 98%