Merck
  • Volatile composition and contribution to the aroma of spanish honeydew honeys. Identification of a new chemical marker.
Volatile composition and contribution to the aroma of spanish honeydew honeys. Identification of a new chemical marker.
Journal of agricultural and food chemistry (2006-06-22)
Lucia Castro-Vázquez, M Consuelo Díaz-Maroto, M Soledad Pérez-Coello
ABSTRACT
Honeydew honeys from holm-oak, oak, and forest were isolated for aroma compounds by simultaneous distillation-extraction and analyzed by gas chromatography-mass spectrometry. In all, 66 volatile components were identified and quantified. trans-Oak lactone, a characteristic volatile component of oak wood, is proposed as a new chemical marker for the plant origin of honeydew honeys. Other compounds, such as aminoacetophenone and propylanisol, could be considered characteristic of holm-oak honeydew honeys. A total of 15 volatile compounds presented odor activity values (OAVs) greater than 1, with phenylacetaldehyde and beta-damascenone being those with the highest OAVs.
Sigma-Aldrich
Damascenone, natural, 1.1-1.4 wt. % (190 proof ethanol), FG
Pricing and availability is not currently available.