Merck
  • The involvement of endogenous opioid mechanisms in the antinociceptive effects induced by antidepressant drugs, desipramine and trimipramine.
The involvement of endogenous opioid mechanisms in the antinociceptive effects induced by antidepressant drugs, desipramine and trimipramine.
Pharmacology, biochemistry, and behavior (2006-05-23)
Yusuf Oztürk, Süleyman Aydin, Rana Beis, Tuba Herekman-Demir
ABSTRACT
The present study was designed to investigate the involvement of endogenous opioid systems in the antinociception induced by the antidepressant drugs, desipramine and trimipramine. For this purpose, the antinociceptive effects of desipramine (7.5 and 15.0 mg/kg i.p.) and trimipramine (5.0 and 10.0 mg/kg i.p.) were compared to that induced by morphine (0.2 and 2.0 mg/kg i.p.) in the tail-clip model in mice. Naloxone (0.3 and 3.0 mg/kg i.p.), a non-specific opioid receptor antagonist, inhibited morphine-induced antinociception in mice, whereas the antinociceptive effects of antidepressant drugs were found to be resistant to naloxone blockade to some extent, since only the higher concentration of naloxone (3.0 mg/kg i.p.) caused significant inhibition of the effects of antidepressant drugs. In contrast, naltrindole (1.0 mg/kg i.p.), a specific delta-receptor antagonist, inhibited antinociception induced by desipramine and trimipramine in this test, while it inhibited the antinociceptive effect of morphine only partly. None of the opioid antagonists produced a significant effect in the tail-clip experiment when they were injected alone. Based on these findings, we concluded that endogenous opioids are involved in the antinociceptive effects of the antidepressant drugs using different mechanisms.
Sigma-Aldrich
Trimipramine maleate salt, ≥98% (TLC), powder