Merck
The molecular basis for wine grape quality--a volatile subject.
Science (New York, N.Y.) (2006-02-14)
Steven T Lund, Joerg Bohlmann
ABSTRACT
Volatile organic compounds are important flavor components of finished wines. In addition to winemaking practices, which shape wine quality, cultivation of the grape berries in the vineyard each season affects the production of volatile organic compounds as well as other chemical components that ultimately contribute to our perception of flavor in finished wines. By studying how berry flavor components are determined by the interplay of vine genotypes, the environment, and cultivation practices at the molecular level, scientists will develop advanced tools and knowledge that will aid viticulturalists in consistently producing balanced, flavorful berries for wine production.
Sigma-Aldrich
2-Isobutyl-3-methoxypyrazine, ≥99%, FG
SKU
Pack Size
Availability
Price
Quantity
Supelco
2-Isobutyl-3-methoxypyrazine solution, certified reference material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Isobutyl-3-methoxypyrazine, 99%