Merck
  • [Synthesis of substituted 3H-indole quaternary ammonium salt and study on its interaction with bovine serum albumin by fluorescence].
[Synthesis of substituted 3H-indole quaternary ammonium salt and study on its interaction with bovine serum albumin by fluorescence].
Guang pu xue yu guang pu fen xi = Guang pu (2005-10-06)
Tong-kuan Xu, Xing-hai Shen, Hong-cheng Gao
ABSTRACT
A substituted 3H-indole quaternary ammonium molecule was designed and synthesized using hexamethylphosphoramide (HMPA) as a solvent. The products were purified and characterized by IR, 1H-NMR, MS and elemental analysis. The binding reaction of this compound with bovine serum albumin (BSA) in aqueous solution was studied using fluorescence. Their binding constant is Ka = 1.995 x 10(5) dm3 x mol(-1) and the binding site number is n = 1.12. It is confirmed that the combination is a single static quenching process.
Sigma-Aldrich
Hexamethylphosphoramide, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexamethylphosphoramide, 99%