Merck
  • Synthesis, characteristics and biological activity of pentacoordinated spirophosphoranes derived from amino acids.
Synthesis, characteristics and biological activity of pentacoordinated spirophosphoranes derived from amino acids.
Amino acids (2005-05-13)
L Yu, Z Liu, H Fang, Q L Zeng, Y F Zhao
ABSTRACT
The reactions of phosphorus trichloride with various amino acids afford the pentacoordinated spirophosphoranes. The reaction procedures were traced by (31)P NMR spectra techniques. A new crystal structure of alanine derivative was characterized, which is a slightly distorted TBP structure. Besides, this kind of spirophosphoranes are potent inhibitors to tyrosinase.
Sigma-Aldrich
Phosphorus trichloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphorus trichloride, ReagentPlus®, 99%
Pricing and availability is not currently available.