Merck
  • Comprehensive two-dimensional liquid chromatography and hyphenated liquid chromatography to study the degradation of poly(bisphenol A)carbonate.
Comprehensive two-dimensional liquid chromatography and hyphenated liquid chromatography to study the degradation of poly(bisphenol A)carbonate.
Journal of chromatography. A (2005-05-03)
L Coulier, E R Kaal, Th Hankemeier
ABSTRACT
Size exclusion chromatography (SEC), gradient polymer elution chromatography (GPEC) and liquid chromatography at critical conditions (LC-CC) have been developed and applied to observe chemical changes in poly(bisphenol A)carbonate (PC) due to hydrolytic degradation. Especially LC-CC appeared to be very successful to observe differences in functionality of PC as result of hydrolytic degradation. Observed differences due to degradation could be identified by (semi) on-line coupling to matrix assisted laser desorption-ionization time-of-flight mass spectrometry (MALDI-TOF-MS). The differences in functionality could be attributed to the formation of different end-groups, i.e. OH end-groups. In addition, comprehensive two-dimensional liquid chromatography (2D-LC) has been applied successfully to study the hydrolytic degradation of PC. LC-CC x SEC showed that the formation of PC with different end-groups occurred over the whole molecular mass range. This information could not be obtained with the separate liquid chromatographic techniques, thereby illustrating the added value of 2D-LC.
Product Description
Supelco
Poly(Bisphenol A carbonate), analytical standard
Sigma-Aldrich
Poly(Bisphenol A carbonate), average Mw ~45,000 by GPC
Supelco
Poly(Bisphenol A carbonate)
Sigma-Aldrich
Poly(Bisphenol A carbonate), melt index 10-12 g/10 min (300°C/1.2kg)