Merck
  • Comparison of cellulose extraction methods for analysis of stable-isotope ratios of carbon and oxygen in plant material.
Comparison of cellulose extraction methods for analysis of stable-isotope ratios of carbon and oxygen in plant material.
Tree physiology (2005-03-03)
Louise E Cullen, Craig Macfarlane
ABSTRACT
The Jayme-Wise and diglyme-HCl methods for extracting cellulose from plant material for stable-isotope analysis differ considerably in ease of use, with the latter requiring significantly less time and specialized equipment. However, the diglyme-HCl method leaves a small lignin residue in the crude cellulose that may affect stable-isotope values, whereas alpha-cellulose produced by the Jayme-Wise method is relatively pure. We examined whether adding a bleaching step to the diglyme-HCl method could produce cellulose of comparable purity to alpha-cellulose by comparing the yield, percent carbon, and carbon (delta13C) and oxygen (delta18O) stable isotope ratios of the two celluloses. We tested each method on the wood of five species that differ in ease of delignification, Eucalyptus maculata Hook., E. botryoides Sm., E. resinifera Sm., Pinus pinaster Ait. and Callitris glaucophylla J. Thompson & L.A.S. Johnson, as well as the foliage of C. glaucophylla. For hardwoods such as the eucalypts, the diglyme-HCl method without bleaching produced cellulose with delta13C and delta18O ratios similar to alpha-cellulose. For the softwood, C. glaucophylla, 3 h of bleaching with acidified chlorite following treatment with diglyme-HCl produced cellulose with delta13C and delta18O ratios similar to alpha-cellulose. Bleached and unbleached crude celluloses and alpha-cellulose of P. pinaster were similar in delta18O, but not delta13C. Both types of crude cellulose produced from the foliage of C. glaucophylla had significantly different isotope ratios from alpha-cellulose. Overall, the diglyme-HCl method, with or without bleaching, appears to be a simple, fast method for extracting alpha-cellulose from hardwoods for stable-isotope analyses, but its suitability for softwoods and foliage needs to be evaluated depending on the species.
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Diethylene glycol dimethyl ether, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Diethylene glycol dimethyl ether, ReagentPlus®, 99%