Merck
  • Mechanisms of arsenite-mediated decreases in benzo[k]fluoranthene-induced human cytochrome P4501A1 levels in HepG2 cells.
Mechanisms of arsenite-mediated decreases in benzo[k]fluoranthene-induced human cytochrome P4501A1 levels in HepG2 cells.
Drug metabolism and disposition: the biological fate of chemicals (2004-12-04)
Erin E Bessette, Michael J Fasco, Brian T Pentecost, Laurence S Kaminsky
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs) and heavy metals are often environmental cocontaminants that could interact to alter PAH carcinogenicity. The heavy metal, arsenite, and the PAH, benzo[k]fluoranthene, were used as prototypes to investigate, in human HepG2 cells, mechanisms whereby the bioactivation of benzo[k]fluoranthene by human CYP1A1 could be diminished by arsenite-mediated decreases in CYP1A1 induction by benzo[k]fluoranthene. To determine whether arsenite down-regulates CYP1A1 transcription, quantitative real-time reverse transcriptase-polymerase chain reaction assays and luciferase reporter gene expression assays were used with HepG2 cells treated with benzo[k]fluoranthene and arsenite, separately and as a mixture. Benzo[k]fluoranthene (0.5 microM) and arsenite (5 microM) markedly decreased benzo[k]fluoranthene-mediated induction of CYP1A1 mRNA by 45%. Plasmids containing the CYP1A1 promoter region (pHu-1A1-FL) were induced 7.4-fold over vehicle by benzo[k]fluoranthene (0.5 microM), whereas arsenite (1, 2.5, or 5 microM) decreased reporter gene expression by 46%, 45%, and 61%, respectively. The plasmid, pHu-1A1-Delta100-FL, lacked xenobiotic response element (XRE) sites at -1061 and -981 and showed greater responsiveness relative to pHu-1A1-FL, by 1.7-fold. Benzo[k]fluoranthene (0.5 microM) and arsenite (1, 2.5, or 5 microM) decreased reporter gene expression by 0%, 27%, and 39%, respectively, relative to expression levels produced by benzo[k]fluoranthene alone. Arsenite is stable for at least 48 h in the HepG2 cell medium with respect to its ability to diminish CYP1A1 benzo[k]fluoranthene induction. Arsenite did not affect benzo[k]fluoranthene induction directly through XRE sites, nor did it affect the stability of CYP1A1 mRNA. Thus, arsenite affects the transcriptional regulation of the benzo[k]fluoranthene-mediated induction of CYP1A1 and could diminish PAH carcinogenicity by decreasing bioactivation by CYP1A1.
Product Description
Supelco
Benzo[k]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%
Pricing and availability is not currently available.
Benzo[k]fluoranthene, BCR®, certified reference material
Pricing and availability is not currently available.